COMH4211BC25 Combi-Blocks, Inc.

5-Amino-1h-indole-3-carboxamide

  • MDL number MFCD05618110
  • Cas Number : 677752-70-4
  • Molecular Weight : 175.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6910

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6910
Availability details for
Quantity
  • Availability
  • Add to Cart