COMH4211BC26 Combi-Blocks, Inc.

Benzyl n-[3-methyl-1-oxo-1-(propan-2-ylamino)pentan-2-yl]carbamate

  • MDL number MFCD02226209
  • Cas Number : 1008457-92-8
  • Molecular Weight : 306.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6911

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6911
Availability details for
Quantity
  • Availability
  • Add to Cart