COMH4211BC27 Combi-Blocks, Inc.

2-Acetyl-5-fluorobenzonitrile

  • MDL number MFCD24713190
  • Cas Number : 1363508-10-4
  • Molecular Weight : 163.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6915

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6915
Availability details for
Quantity
  • Availability
  • Add to Cart