COMH4211BC28 Combi-Blocks, Inc.

3-Bromo-n-(cyclopropylmethyl)-4-methylaniline

  • MDL number MFCD16321048
  • Cas Number : 1226261-56-8
  • Molecular Weight : 240.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-N-(cyclopropylmethyl)-4-methylaniline
Inchi Code 1S/C11H14BrN/c1-8-2-5-10(6-11(8)12)13-7-9-3-4-9/h2,5-6,9,13H,3-4,7H2,1H3
Inchi Key YVXYUAPZLIUFFX-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6917

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6917
Availability details for
Quantity
  • Availability
  • Add to Cart