COMH4211BC2A Combi-Blocks, Inc.

6-Amino-n,n-dimethylpyridine-3-carboxamide

  • MDL number MFCD16688690
  • Cas Number : 827588-33-0
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6920

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6920
Availability details for
Quantity
  • Availability
  • Add to Cart