COMH4211BC2C Combi-Blocks, Inc.

Methyl 2-chloro-5-nitro-4-(propan-2-yloxy)benzoate

  • MDL number MFCD30607336
  • Cas Number : 1143026-86-1
  • Molecular Weight : 273.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6923

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6923
Availability details for
Quantity
  • Availability
  • Add to Cart