COMH4211BC2E Combi-Blocks, Inc.

4-(Ethylsulfonyl)-3-methylbenzoic acid

  • MDL number MFCD30290509
  • Cas Number : 1392466-96-4
  • Molecular Weight : 228.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6927

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6927
Availability details for
Quantity
  • Availability
  • Add to Cart