COMH4211BC30 Combi-Blocks, Inc.

N-(2-(2-(2-Aminoethoxyl)ethoxy)ethyl)-n-methyl-2-nitrobenzenesulfonamide

  • MDL number MFCD31579561
  • Cas Number : 1571191-79-1
  • Molecular Weight : 347.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(2-(2-(2-aminoethoxy)ethoxy)ethyl)-N-methyl-2-nitrobenzenesulfonamide
Inchi Code 1S/C13H21N3O6S/c1-15(7-9-22-11-10-21-8-6-14)23(19,20)13-5-3-2-4-12(13)16(17)18/h2-5H,6-11,14H2,1H3
Inchi Key YUFPCRWYTACWGQ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6931

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6931
Availability details for
Quantity
  • Availability
  • Add to Cart