COMH4211BC31 Combi-Blocks, Inc.

2-(2-Methyl-2h-1,2,3,4-tetrazol-5-yl)benzaldehyde

  • MDL number MFCD22045708
  • Cas Number : 205452-87-5
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6932

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6932
Availability details for
Quantity
  • Availability
  • Add to Cart