COMH4211BC33 Combi-Blocks, Inc.

N-Cyclopropylmethyl-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-benzamide

  • MDL number MFCD30742727
  • Cas Number : 515135-62-3
  • Molecular Weight : 301.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6935

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6935
Availability details for
Quantity
  • Availability
  • Add to Cart