COMH4211BC34 Combi-Blocks, Inc.

4,5-Dichloro-2-cyclopropylpyrimidine

  • MDL number MFCD16987925
  • Cas Number : 1240595-61-2
  • Molecular Weight : 189.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6940

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6940
Availability details for
Quantity
  • Availability
  • Add to Cart