COMH4211BC35 Combi-Blocks, Inc.

(1-Ethanesulfonyl-piperidin-4-yl)-acetic acid

  • MDL number MFCD11585619
  • Cas Number : 1030420-97-3
  • Molecular Weight : 235.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6942

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6942
Availability details for
Quantity
  • Availability
  • Add to Cart