COMH4211BC36 Combi-Blocks, Inc.

2-[4-(2-Fluoro-benzyloxy)-phenyl]-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane

  • MDL number MFCD31583129
  • Cas Number : 2246642-41-9
  • Molecular Weight : 328.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6947

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6947
Availability details for
Quantity
  • Availability
  • Add to Cart