COMH4211BC37 Combi-Blocks, Inc.

3-Cyclopropylmethylsulfanyl-propionic acid

  • MDL number MFCD12138602
  • Cas Number : 1154384-42-5
  • Molecular Weight : 160.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6950

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6950
Availability details for
Quantity
  • Availability
  • Add to Cart