COMH4211BC39 Combi-Blocks, Inc.

4-[(4-Methoxyphenyl)methoxy]iodobenzene

  • MDL number MFCD18978182
  • Cas Number : 682756-44-1
  • Molecular Weight : 340.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6953

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6953
Availability details for
Quantity
  • Availability
  • Add to Cart