COMH4211BC3C Combi-Blocks, Inc.

4,4,5,5-Tetramethyl-2-[4-(1-phenyl-ethoxy)-phenyl]-[1,3,2]dioxaborolane

  • MDL number MFCD31583132
  • Molecular Weight : 324.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6958

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6958
Availability details for
Quantity
  • Availability
  • Add to Cart