COMH4211BC3E Combi-Blocks, Inc.

4-Bromo-n-[(2r)-1-hydroxypropan-2-yl]-3-methylbenzamide

  • MDL number MFCD26856317
  • Cas Number : 1568166-53-9
  • Molecular Weight : 272.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6961

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6961
Availability details for
Quantity
  • Availability
  • Add to Cart