COMH4211BC3F Combi-Blocks, Inc.

N-[2-Fluoro-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]cyclopropanecarboxamide

  • MDL number MFCD31583242
  • Molecular Weight : 305.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6965

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6965
Availability details for
Quantity
  • Availability
  • Add to Cart