COMH4211BC44 Combi-Blocks, Inc.

2-[1-(2-Methylpropyl)piperidin-4-yl]ethan-1-ol

  • MDL number MFCD23708532
  • Cas Number : 1427375-89-0
  • Molecular Weight : 185.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6973

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6973
Availability details for
Quantity
  • Availability
  • Add to Cart