COMH4211BC45 Combi-Blocks, Inc.

3-(Methoxymethoxy)-n-methylbenzylamine

  • MDL number MFCD21195278
  • Cas Number : 475086-51-2
  • Molecular Weight : 181.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6975

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6975
Availability details for
Quantity
  • Availability
  • Add to Cart