COMH4211BC48 Combi-Blocks, Inc.

2-(4-Chloro-phenoxy)-1-piperazin-1-yl-ethanone

  • MDL number MFCD03446007
  • Cas Number : 143999-83-1
  • Molecular Weight : 254.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6980

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6980
Availability details for
Quantity
  • Availability
  • Add to Cart