COMH4211BC4D Combi-Blocks, Inc.

Ethyl 4-amino-3-trifluoromethyl-phenoxy-acetate

  • MDL number MFCD31579350
  • Cas Number : 751463-15-7
  • Molecular Weight : 263.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6992

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6992
Availability details for
Quantity
  • Availability
  • Add to Cart