COMH4211BC50 Combi-Blocks, Inc.

4-Bromo-3-(3-methoxypropoxy)benzoic acid

  • MDL number MFCD25954808
  • Cas Number : 1044563-86-1
  • Molecular Weight : 289.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6997

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6997
Availability details for
Quantity
  • Availability
  • Add to Cart