COMH4211BC51 Combi-Blocks, Inc.

Cyclohexyl (4-nitrophenyl) carbonate

  • MDL number MFCD30185827
  • Cas Number : 176787-42-1
  • Molecular Weight : 265.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6998

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6998
Availability details for
Quantity
  • Availability
  • Add to Cart