COMH4211BC52 Combi-Blocks, Inc.

2-[(2,4-Dimethylphenyl)formamido]propanoic acid

  • MDL number MFCD09836860
  • Cas Number : 1117700-39-6
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(2,4-dimethylbenzoyl)alanine
Inchi Code 1S/C12H15NO3/c1-7-4-5-10(8(2)6-7)11(14)13-9(3)12(15)16/h4-6,9H,1-3H3,(H,13,14)(H,15,16)
Inchi Key SZNNWUTYNAYBQU-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7000

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7000
Availability details for
Quantity
  • Availability
  • Add to Cart