COMH4211BC57 Combi-Blocks, Inc.

2-(Bromomethyl)-4-iodo-1-methylbenzene

  • MDL number MFCD16251115
  • Cas Number : 1261647-64-6
  • Molecular Weight : 311.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7010

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7010
Availability details for
Quantity
  • Availability
  • Add to Cart