COMH4211BC58 Combi-Blocks, Inc.

3-Amino-5-isopropoxybenzamide

  • MDL number MFCD29098621
  • Cas Number : 1369941-62-7
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7013

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7013
Availability details for
Quantity
  • Availability
  • Add to Cart