COMH4211BC59 Combi-Blocks, Inc.

3-(2,3-Difluorophenyl)-2-methylpropanoic acid

  • MDL number MFCD12794509
  • Cas Number : 1250203-60-1
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7023

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7023
Availability details for
Quantity
  • Availability
  • Add to Cart