COMH4211BC5B Combi-Blocks, Inc.

2-Chloro-6-iodomethyl-pyridine

  • MDL number MFCD28556944
  • Cas Number : 1935998-77-8
  • Molecular Weight : 253.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7033

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7033
Availability details for
Quantity
  • Availability
  • Add to Cart