COMH4211BC5C Combi-Blocks, Inc.

2-Iodo-1-isopropoxy-4-(trifluoromethyl)benzene

  • MDL number MFCD29095490
  • Cas Number : 1369872-66-1
  • Molecular Weight : 330.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7035

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7035
Availability details for
Quantity
  • Availability
  • Add to Cart