COMH4211BCAA Combi-Blocks, Inc.

3'-Formyl-4'-methoxy-2-(trifluoromethyl)biphenyl-4-carbonitrile

  • MDL number MFCD31583175
  • Cas Number : 923281-61-2
  • Molecular Weight : 305.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7039

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7039
Availability details for
Quantity
  • Availability
  • Add to Cart