COMH4211BCAB Combi-Blocks, Inc.

2-Chloro-3-(2-methylpropoxy)pyridine

  • MDL number MFCD21106857
  • Cas Number : 1486099-41-5
  • Molecular Weight : 185.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7041

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7041
Availability details for
Quantity
  • Availability
  • Add to Cart