COMH4211BCAD Combi-Blocks, Inc.

Trans (4-amino-cyclohexyl)-piperidin-1-yl-methanone

  • MDL number MFCD31557886
  • Cas Number : 412291-07-7
  • Molecular Weight : 210.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7044

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7044
Availability details for
Quantity
  • Availability
  • Add to Cart