COMH4211BCAE Combi-Blocks, Inc.

(3-Chloro-4-propoxybenzyl)amine

  • MDL number MFCD14656255
  • Cas Number : 776997-51-4
  • Molecular Weight : 199.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7047

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7047
Availability details for
Quantity
  • Availability
  • Add to Cart