COMH4211BF62 Combi-Blocks, Inc.

Ethyl 2-hydroxy-5-nitrobenzoate

  • MDL number MFCD01365750
  • Cas Number : 42348-40-3
  • Molecular Weight : 211.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7068

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7068
Availability details for
Quantity
  • Availability
  • Add to Cart