COMH4211BF64 Combi-Blocks, Inc.

3-(4-Chlorophenyl)-4-isoxazolecarboxylic acid ethyl ester

  • MDL number MFCD00981158
  • Cas Number : 29363-98-2
  • Molecular Weight : 251.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7075

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7075
Availability details for
Quantity
  • Availability
  • Add to Cart