COMH4211BF66 Combi-Blocks, Inc.

4-Difluoromethoxy-3-methoxybenzylamine hydrochloride

  • MDL number MFCD25370995
  • Cas Number : 1431966-33-4
  • Molecular Weight : 239.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7078

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7078
Availability details for
Quantity
  • Availability
  • Add to Cart