COMH4211BF69 Combi-Blocks, Inc.

1-(2-(2-Bromoethoxy)-5-fluorophenyl)ethan-1-one

  • MDL number MFCD23825363
  • Cas Number : 1282530-48-6
  • Molecular Weight : 261.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-(2-bromoethoxy)-5-fluorophenyl)ethan-1-one
Inchi Code 1S/C10H10BrFO2/c1-7(13)9-6-8(12)2-3-10(9)14-5-4-11/h2-3,6H,4-5H2,1H3
Inchi Key ZEUQGAIAUMESIN-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7083

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7083
Availability details for
Quantity
  • Availability
  • Add to Cart