COMH4211BF6B Combi-Blocks, Inc.

3-Fluoro-4-hydroxy-n,n-dimethylbenzamide

  • MDL number MFCD16997908
  • Cas Number : 1236284-45-9
  • Molecular Weight : 183.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7085

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7085
Availability details for
Quantity
  • Availability
  • Add to Cart