COMH4211BF6D Combi-Blocks, Inc.

3-(5-Fluoro-2-methylphenyl)propan-1-ol

  • MDL number MFCD09926865
  • Cas Number : 1000551-43-8
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7090

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7090
Availability details for
Quantity
  • Availability
  • Add to Cart