COMH4211BF6E Combi-Blocks, Inc.

(R)-2-Amino-3-methyl-1-(pyrrolidin-1-yl)butan-1-one

  • MDL number MFCD20486003
  • Cas Number : 1334701-05-1
  • Molecular Weight : 170.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7093

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7093
Availability details for
Quantity
  • Availability
  • Add to Cart