COMH4211BF71 Combi-Blocks, Inc.

5-Bromo-2-(4-fluoro-phenylsulfanyl)-benzonitrile

  • MDL number MFCD31381721
  • Cas Number : 1778487-87-8
  • Molecular Weight : 308.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7097

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7097
Availability details for
Quantity
  • Availability
  • Add to Cart