COMH4211BF74 Combi-Blocks, Inc.

2-(6-Hydroxy-naphthalen-2-yl)-propionic acid methyl ester

  • MDL number MFCD00100505
  • Cas Number : 61495-42-9
  • Molecular Weight : 230.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7102

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7102
Availability details for
Quantity
  • Availability
  • Add to Cart