COMH4211BF75 Combi-Blocks, Inc.

4-(3,4-Dichloro-phenylsulfanyl)-butylamine

  • MDL number MFCD11585861
  • Cas Number : 1082501-54-9
  • Molecular Weight : 250.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7105

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7105
Availability details for
Quantity
  • Availability
  • Add to Cart