COMH4211BF76 Combi-Blocks, Inc.

5-Bromo-1-ethyl-1,3-dihydro-benzoimidazol-2-one

  • MDL number MFCD11868740
  • Cas Number : 334829-57-1
  • Molecular Weight : 241.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7107

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7107
Availability details for
Quantity
  • Availability
  • Add to Cart