COMH4211BF77 Combi-Blocks, Inc.

1-(2-Fluoro-5-(trifluoromethyl)phenyl)ethanamine hydrochloride

  • MDL number MFCD30725582
  • Cas Number : 1373865-26-9
  • Molecular Weight : 243.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7108

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7108
Availability details for
Quantity
  • Availability
  • Add to Cart