COMH4211BF78 Combi-Blocks, Inc.

5-Fluoro-1-propyl-1h-indole-2-carboxylic acid

  • MDL number MFCD23161937
  • Cas Number : 1391467-31-4
  • Molecular Weight : 221.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7112

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7112
Availability details for
Quantity
  • Availability
  • Add to Cart