COMH4211BF79 Combi-Blocks, Inc.

1-(4-Bromo-2-fluorophenyl)-2,2,2-trifluoroethanol

  • MDL number MFCD16067945
  • Cas Number : 1033805-88-7
  • Molecular Weight : 273.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7114

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7114
Availability details for
Quantity
  • Availability
  • Add to Cart