COMH4211BF7B Combi-Blocks, Inc.

[(2-Bromo-5-fluorophenyl)methyl](methyl)amine

  • MDL number MFCD09807033
  • Cas Number : 887139-05-1
  • Molecular Weight : 218.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7120

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7120
Availability details for
Quantity
  • Availability
  • Add to Cart