COMH4211BF7D Combi-Blocks, Inc.

1-(2,4-Difluoro-benzyl)-3,5-dimethyl-1h-pyrazol-4-ol

  • MDL number MFCD31583225
  • Cas Number : 2270905-98-9
  • Molecular Weight : 238.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7123

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-7123
Availability details for
Quantity
  • Availability
  • Add to Cart